(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol 97%

Supplier: Ambeed
Warning

A432202-1G A432202-250MG
76648-950EA 334.19 USD
76648-950 76648-952
(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol 97%
(2-Amino-5-(3-(piperidin-1-yl)propoxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O₂
MW: 264.3633 g/mol
MDL Number: MFCD31715177
CAS Number: 2288710-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR