1-(5,6,7,8-Tetrahydronaphthalen-2-yl)pentan-1-one 95%

Supplier: Ambeed
Warning

A432073-100MG A432073-1G A432073-5G
76648-944EA 20.16 USD
76648-944 76648-946 76648-948
1-(5,6,7,8-Tetrahydronaphthalen-2-yl)pentan-1-one 95%
1-(5,6,7,8-Tetrahydronaphthalen-2-yl)pentan-1-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀O
MW: 216.3187 g/mol
MDL Number: MFCD11105512
CAS Number: 101498-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR