1-tert-Butyl 6-methyl 3-bromo-1H-indole-1,6-dicarboxylate 95%

Supplier: Ambeed
Warning

A431318-1G A431318-250MG A431318-5G
76648-938EA 107.16 USD
76648-938 76648-940 76648-942
1-tert-Butyl 6-methyl 3-bromo-1H-indole-1,6-dicarboxylate 95%
1-tert-Butyl 6-methyl 3-bromo-1H-indole-1,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₆BrNO₄
MW: 354.1958 g/mol
MDL Number: MFCD30067555
CAS Number: 479552-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR