2,4-Dibromo-6-nitroquinoline 97%

Supplier: Ambeed
Warning

A1349538-100MG A1349538-1G A1349538-250MG A1349538-5G
76658-074EA 64.72 USD
76658-074 76658-076 76658-078 76658-080
2,4-Dibromo-6-nitroquinoline 97%
2,4-Dibromo-6-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₄Br₂N₂O₂
MW: 331.9483 g/mol
MDL Number: MFCD18254547
CAS Number: 437708-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR