4-(N-(4-Cyclohexylbenzyl)-2-(2,3,4,5,6-pentafluoro-N-methylphenylsulfonamido)acetamido)-N-hydroxybenzamide 97%

Supplier: Ambeed
Warning

Synonyms: SH5-07

A427567-100MG A427567-10MG A427567-1MG A427567-50MG A427567-5MG
76648-900EA 281.14 USD
76648-900 76648-902 76648-904 76648-906 76648-908
4-(N-(4-Cyclohexylbenzyl)-2-(2,3,4,5,6-pentafluoro-N-methylphenylsulfonamido)acetamido)-N-hydroxybenzamide 97%
4-(N-(4-Cyclohexylbenzyl)-2-(2,3,4,5,6-pentafluoro-N-methylphenylsulfonamido)acetamido)-N-hydroxybenzamide

Precaución: for Research Use Only

Formula: C₂₉H₂₈F₅N₃O₅S
MW: 625.61 g/mol
MDL Number: MFCD30718177
CAS Number: 1456632-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR