Ethyl 1H-pyrazole-4-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A423962-1G A423962-25G A423962-5G
76648-888EA 11 USD
76648-888 76648-890 76648-892
Ethyl 1H-pyrazole-4-carboxylate hydrochloride 98%
Ethyl 1H-pyrazole-4-carboxylate hydrochloride
Formula: C₆H₉ClN₂O₂
MDL Number: MFCD16038569
CAS Number: 1242339-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR