tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate 98%

Supplier: Ambeed
Warning

A421256-100MG A421256-1G A421256-250MG
76648-876EA 36.08 USD
76648-876 76648-878 76648-880
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate 98%
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₅BN₂O₄
MW: 344.2131 g/mol
MDL Number: MFCD22577263
CAS Number: 1446443-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR