[Dicyclohexyl[3-(1-methylethoxy)-2′,4′,6′-tris(1-methylethyl)[1,1′-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium 98%

Supplier: Ambeed
Warning

A417851-250MG A417851-1G A417851-100MG A417851-5G
76648-874EA 201.58 USD
76648-874 77551-264 77616-812 77748-845
[Dicyclohexyl[3-(1-methylethoxy)-2′,4′,6′-tris(1-methylethyl)[1,1′-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium 98%
[Dicyclohexyl[3-(1-methylethoxy)-2′,4′,6′-tris(1-methylethyl)[1,1′-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)[2′-(methylamino-κN)[1,1′-biphenyl]-2-yl-κC]palladium

Precaución: for Research Use Only

Formula: C₅₀H₇₀NO₄PPdS
MW: 918.55 g/mol
CAS Number: 2132978-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR