4H-1,2,4-Triazol-3-amine 95%

Supplier: Ambeed
Warning

A417316-500G A417316-100G A417316-25G
76648-872EA 96.55 USD
76648-872 77572-302 77594-290
4H-1,2,4-Triazol-3-amine 95%
4H-1,2,4-Triazol-3-amine

Precaución: for Research Use Only

Formula: C₂H₄N₄
MW: 84.0800 g/mol
MDL Number: MFCD00053362
CAS Number: 65312-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR