Hydroquinidine 1,4-phthalazinediyl ether mixture (DHQD)2PHAL:0.552% ≥98%

Supplier: Ambeed
Warning

Synonyms: AD-mix-β

A416864-10G A416864-1G A416864-25G A416864-500G A416864-5G A416864-100G
76648-860EA 37.14 USD
76648-860 76648-862 76648-864 76648-866 76648-868 77551-252
Hydroquinidine 1,4-phthalazinediyl ether mixture (DHQD)2PHAL:0.552% ≥98%
Hydroquinidine 1,4-phthalazinediyl ether mixture

Precaución: for Research Use Only

Formula: C₄₈H₅₄N₆O₄
MW: 778.98 g/mol
MDL Number: MFCD00198107
CAS Number: 148618-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR