EX-229 95%

Supplier: Ambeed
Warning

A414474-10MG A414474-1MG A414474-25MG A414474-5MG
76648-846EA 56 USD
76648-846 76648-848 76648-850 76648-852
EX-229 95%
EX-229

Precaución: for Research Use Only

Formula: C₂₄H₁₈ClN₃O₃
MDL Number: MFCD31696597
CAS Number: 1219739-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR