(6-(Methoxycarbonyl)naphthalen-2-yl)boronic acid 95%
Supplier: Ambeed
|
Warning
|
A413144-100MG
A413144-1G
A413144-250MG
A413144-5G
76648-840EA
10
USD
76648-840
76648-842
76648-844
77748-836
(6-(Methoxycarbonyl)naphthalen-2-yl)boronic acid 95%
(6-(Methoxycarbonyl)naphthalen-2-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₁BO₄ MW: 230.0243 g/mol |
MDL Number:
MFCD32065669 CAS Number: 1809899-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...