[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyrromethene 546, 5,5-Difluoro-1,3,7,9,10-pentamethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide, BODIPY 493/503
A411914-100MG
A411914-1G
A411914-250MG
A411914-5G
76648-836EA
31.83
USD
76648-836
77616-790
77616-792
77748-832
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane) 99%
[[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]methane](difluoroborane)
Precaución: for Research Use Only
Formula:
C₁₄H₁₇BF₂N₂ MW: 262.11 g/mol |
MDL Number:
MFCD00467372 CAS Number: 121207-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...