2-Amino-5-(difluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A411284-100MG A411284-1G A411284-250MG
76648-828EA 122.01 USD
76648-828 76648-830 76648-832
2-Amino-5-(difluoromethoxy)benzoic acid 97%
2-Amino-5-(difluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₃
MW: 203.1429 g/mol
MDL Number: MFCD29493186
CAS Number: 1108666-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR