4-Bromo-2H-benzo[d][1,2,3]triazole 95%

Supplier: Ambeed
Danger

A410465-100MG A410465-250MG A410465-1G
76648-816EA 48.81 USD
76648-816 76648-818 77594-250
4-Bromo-2H-benzo[d][1,2,3]triazole 95%
4-Bromo-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
MDL Number: MFCD31640987
CAS Number: 2058157-38-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR