Methyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate 98%

Supplier: Ambeed
Warning

A408204-100MG A408204-1G A408204-250MG
76648-810EA 205.82 USD
76648-810 76648-812 76648-814
Methyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate 98%
Methyl 3-iodobicyclo[1.1.1]pentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉IO₂
MW: 252.0496 g/mol
MDL Number: MFCD30802630
CAS Number: 141046-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR