3-(Aminomethyl)bicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride 97%

Supplier: Ambeed
Warning

A405760-100MG A405760-50MG A405760-250MG
76648-786EA 218.55 USD
76648-786 76648-788 77748-819
3-(Aminomethyl)bicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride 97%
3-(Aminomethyl)bicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂ClNO₂
MW: 177.6287 g/mol
MDL Number: MFCD31617921
CAS Number: 2173991-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR