(Triphenylphosphonio)difluoroacetate 95%

Supplier: Ambeed
Warning

Synonyms: 2,2-Difluoro-2-(triphenylphosphonio)acetate

A403478-100MG A403478-1G A403478-250MG A403478-5G A403478-25G
76648-778EA 14.86 USD
76648-778 76648-780 76648-782 76648-784 77568-816
(Triphenylphosphonio)difluoroacetate 95%
(Triphenylphosphonio)difluoroacetate

Precaución: For research use only.

Formula: C₂₀H₁₅F₂O₂P
MW: 356.302 g/mol
MDL Number: MFCD28359690
CAS Number: 1449521-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR