3,3'-Bi-9H-carbazole,9-[1'1-biphenyl]-3-yl-9'-phenyl 98%

Supplier: Ambeed
Warning

A401674-1G A401674-250MG
76648-768EA 70.02 USD
76648-768 76648-770
3,3'-Bi-9H-carbazole,9-[1'1-biphenyl]-3-yl-9'-phenyl 98%
3,3'-Bi-9H-carbazole,9-[1'1-biphenyl]-3-yl-9'-phenyl

Precaución: for Research Use Only

Formula: C₄₂H₂₈N₂
MW: 560.6851 g/mol
MDL Number: MFCD32664643
CAS Number: 1619966-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR