2-Bromo-1-(3,3-dinitroazetidin-1-yl)ethan-1-one 95%

Supplier: Ambeed
Danger

A400874-10MG A400874-1MG A400874-5MG
76648-760EA 43 USD
76648-760 76648-762 76648-764 76648-766
2-Bromo-1-(3,3-dinitroazetidin-1-yl)ethan-1-one 95%
2-Bromo-1-(3,3-dinitroazetidin-1-yl)ethan-1-one

Precaución: for Research Use Only

Formula: C₅H₆BrN₃O₅
MDL Number: MFCD25976849
CAS Number: 925206-65-1
UN: 2926
ADR: 4.1,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR