Ethyl 6-bromo-4-chloro-7-fluoroquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A400431-1G A400431-5G
76648-756EA 218.55 USD
76648-756 76648-758
Ethyl 6-bromo-4-chloro-7-fluoroquinoline-3-carboxylate 97%
Ethyl 6-bromo-4-chloro-7-fluoroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₈BrClFNO₂
MW: 332.5528 g/mol
MDL Number: MFCD26333951
CAS Number: 953803-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR