4,4,5,5-Tetramethyl-2-(selenophen-2-yl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Danger

A397367-1G A397367-250MG
76648-746EA 330 USD
76648-746 76648-748
4,4,5,5-Tetramethyl-2-(selenophen-2-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(selenophen-2-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₂Se
MDL Number: MFCD28969360
CAS Number: 1338368-18-5
UN: 3283
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR