5-(tert-Butyl)-1,3-oxazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 5-tert-butyl-1,3-oxazole-4-carboxylic acid

A396782-100MG A396782-1G A396782-250MG
76648-740EA 51 USD
76648-740 76648-742 76648-744
5-(tert-Butyl)-1,3-oxazole-4-carboxylic acid 97%
5-(tert-Butyl)-1,3-oxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃
MDL Number: MFCD07376154
CAS Number: 914637-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR