(S)-3-Chloro-7-(2-(hydroxymethyl)morpholino)-2-methyl-5-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-6-carbonitrile 98+%

Supplier: Ambeed
Warning

A393036-10MG A393036-1MG A393036-25MG A393036-50MG A393036-5MG
76648-724EA 269.47 USD
76648-724 76648-726 76648-728 76648-730 76648-732
(S)-3-Chloro-7-(2-(hydroxymethyl)morpholino)-2-methyl-5-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-6-carbonitrile 98+%
(S)-3-Chloro-7-(2-(hydroxymethyl)morpholino)-2-methyl-5-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₄ClF₃N₄O₂
MW: 374.7455 g/mol
MDL Number: MFCD31657405
CAS Number: 2227425-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR