1-Acetylnaphthalene 95%

Supplier: Ambeed
Warning

Synonyms: 1'-Acetonaphthone, α-acetonaphthone, 1'-Acetylnaphthalene, Methyl α-naphthyl ketone, Methyl 1-naphthyl ketone, 1-Acetonaphthone

A391261-100G A391261-10G A391261-25G A391261-500G
76648-716EA 11.67 USD
76648-716 76648-718 76648-720 76648-722
1-Acetylnaphthalene 95%
1-Acetylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
MDL Number: MFCD00004013
CAS Number: 941-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR