(E)-(3-Chlorostyryl)boronic acid 97%

Supplier: Ambeed
Warning

A389845-1G A389845-250MG A389845-5G
76648-702EA 216 USD
76648-702 76648-704 76648-706
(E)-(3-Chlorostyryl)boronic acid 97%
(E)-(3-Chlorostyryl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BClO₂
MW: 182.41 g/mol
CAS Number: 1258237-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR