tert-Butyl 14-(tosyloxy)-3,6,9,12-tetraoxatetradecan-1-oate 95%

Supplier: Ambeed
Warning

Synonyms: Tos-PEG4-CH2-Boc

A388377-100MG A388377-1G A388377-250MG A388377-5G
76648-696EA 29.71 USD
76648-696 76648-698 76648-700 77748-783
tert-Butyl 14-(tosyloxy)-3,6,9,12-tetraoxatetradecan-1-oate 95%
tert-Butyl 14-(tosyloxy)-3,6,9,12-tetraoxatetradecan-1-oate

Precaución: for Research Use Only

Formula: C₂₁H₃₄O₉S
MW: 462.5543 g/mol
MDL Number: MFCD27977499
CAS Number: 169751-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR