(S)-Oxetan-2-ylmethanamine ≥97%, ee 99%

Supplier: Ambeed
Warning

A387383-100MG A387383-1G A387383-250MG A387383-50MG A387383-5G
76648-684EA 23 USD
76648-684 76648-686 76648-688 76648-690 77572-162
(S)-Oxetan-2-ylmethanamine ≥97%, ee 99%
(S)-Oxetan-2-ylmethanamine

Precaución: for Research Use Only

Formula: C₄H₉NO
MDL Number: MFCD31727897
CAS Number: 2091328-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR