(E)-1-(2-(Methylthio)-1-(2-(pentyloxy)phenyl)vinyl)-1H-imidazole hydrochloride 99+%
Supplier: Ambeed
|
Warning
|
A386908-1MG
A386908-5MG
76648-678EA
11
USD
76648-678
76648-680
(E)-1-(2-(Methylthio)-1-(2-(pentyloxy)phenyl)vinyl)-1H-imidazole hydrochloride 99+%
(E)-1-(2-(Methylthio)-1-(2-(pentyloxy)phenyl)vinyl)-1H-imidazole hydrochloride
Precaución: for Research Use Only
Formula:
C₁₇H₂₃ClN₂OS MW: 338.9 g/mol |
MDL Number:
MFCD00895755 CAS Number: 130773-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...