4-Bromoquinoline-6-carboxamide 95%

Supplier: Ambeed
Warning

A386160-100MG A386160-250MG
76648-674EA 71 USD
76648-674 76648-676
4-Bromoquinoline-6-carboxamide 95%
4-Bromoquinoline-6-carboxamide

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂O
MDL Number: MFCD31698989
CAS Number: 2383346-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR