R-7050 97%

Supplier: Ambeed
Warning

Synonyms: 8-Chloro-4-phenylsulphanyl-1-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]quinoxaline, TNF-alpha Antagonist III, 8-Chloro-1-(trifluoromethyl)[1,2,4]triazolo[4,3-a]quinoxalin-4-yl phenyl sulphide, 8-Chloro-4-(phenylthio)-1-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]quinoxaline

A386124-100MG A386124-10MG A386124-1MG A386124-25MG A386124-50MG A386124-5MG
76648-662EA 203.7 USD
76648-662 76648-664 76648-666 76648-668 76648-670 76648-672
R-7050 97%
R-7050

Precaución: for Research Use Only

Formula: C₁₆H₈ClF₃N₄S
MW: 380.77 g/mol
MDL Number: MFCD00140737
CAS Number: 303997-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR