2-(3-(3-(Trifluoromethyl)phenyl)ureido)benzoic acid 97%

Supplier: Ambeed
Warning

A385750-10MG A385750-1MG A385750-5MG
76648-656EA 658.82 USD
76648-656 76648-658 76648-660
2-(3-(3-(Trifluoromethyl)phenyl)ureido)benzoic acid 97%
2-(3-(3-(Trifluoromethyl)phenyl)ureido)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃N₂O₃
MW: 324.2547 g/mol
MDL Number: MFCD00248828
CAS Number: 1566-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR