Methyl 6-bromo-4-methoxypicolinate 95%

Supplier: Ambeed
Warning

A382723-100MG A382723-250MG
76648-644EA 113.52 USD
76648-644 76648-646
Methyl 6-bromo-4-methoxypicolinate 95%
Methyl 6-bromo-4-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.0580 g/mol
MDL Number: MFCD18257514
CAS Number: 1256803-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR