N-(4,6-Dimethylpyridin-2-yl)-4-(4-(trifluoromethyl)benzyl)piperazine-1-carbothioamide 97%

Supplier: Ambeed
Warning

A380274-10MG A380274-1MG A380274-25MG A380274-5MG A380274-50MG
76648-628EA 78 USD
76648-628 76648-630 76648-632 76648-634 77622-296
N-(4,6-Dimethylpyridin-2-yl)-4-(4-(trifluoromethyl)benzyl)piperazine-1-carbothioamide 97%
N-(4,6-Dimethylpyridin-2-yl)-4-(4-(trifluoromethyl)benzyl)piperazine-1-carbothioamide

Precaución: for Research Use Only

Formula: C₂₀H₂₃F₃N₄S
MDL Number: MFCD30343875
CAS Number: 1916571-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR