4-Hydroxy-6-methylpyridin-2(3H)-one 95%

Supplier: Ambeed
Warning

A377504-100G A377504-1G A377504-500G A377504-5G A377504-25G
76648-620EA 176.11 USD
76648-620 76648-622 76648-624 76648-626 77616-652
4-Hydroxy-6-methylpyridin-2(3H)-one 95%
4-Hydroxy-6-methylpyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.13 g/mol
MDL Number: MFCD01860849
CAS Number: 70254-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR