dibenzo[b,d]furan-3-ylboronic acid 98%

Supplier: Ambeed
Warning

A376828-100MG A376828-250MG A376828-25G A376828-5G A376828-1G
76648-612EA 5.31 USD
76648-612 76648-614 76648-616 76648-618 77748-755
dibenzo[b,d]furan-3-ylboronic acid 98%
dibenzo[b,d]furan-3-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₉BO₃
MW: 212.0091 g/mol
MDL Number: MFCD26516274
CAS Number: 395087-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR