Methyl 3-hydroxybicyclo[1.1.1]pentane-1-carboxylate 98%

Supplier: Ambeed
Warning

A371949-100MG A371949-1G A371949-250MG A371949-50MG
76648-602EA 241.89 USD
76648-602 76648-604 76648-606 76648-608
Methyl 3-hydroxybicyclo[1.1.1]pentane-1-carboxylate 98%
Methyl 3-hydroxybicyclo[1.1.1]pentane-1-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.1525 g/mol
MDL Number: MFCD30802629
CAS Number: 2092825-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR