3-(Di-tert-butylphosphonium)propane sulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(Di-tert-butylphosphino)propane-1-sulphonic acid
A371781-10G
A371781-5G
76648-598EA
323.58
USD
76648-598
76648-600
3-(Di-tert-butylphosphonium)propane sulfonate 98%
3-(Di-tert-butylphosphonium)propane sulfonate
Precaución: for Research Use Only
Formula:
C₁₁H₂₅O₃PS MW: 268.35 g/mol |
MDL Number:
MFCD12546062 CAS Number: 1055888-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...