E-7046 99%

Supplier: Ambeed
Warning

Synonyms: Palupiprant, 4-[(1S)-1-[[[3-(difluoromethyl)-1-methyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazol-4-yl]carbonyl]amino]ethyl]-benzoic acid, (S)-4-(1-(3-(Difluoromethyl)-1-methyl-5-(3-(trifluoromethyl)phenoxy)-1H-pyrazole-4-carboxamido)ethyl)benzoic acid

A368414-10MG A368414-1G A368414-1MG A368414-250MG A368414-25MG A368414-50MG A368414-5MG A368414-100MG
76648-582EA 55.17 USD
76648-582 76648-584 76648-586 76648-588 76648-590 76648-592 76648-594 77748-734
E-7046 99%
E-7046

Precaución: for Research Use Only

Formula: C₂₂H₁₈F₅N₃O₄
MW: 483.41 g/mol
MDL Number: MFCD30502665
CAS Number: 1369489-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR