4-(3-Hydroxyazetidin-1-yl)benzoic acid 95%

Supplier: Ambeed
Warning

A366749-100MG A366749-1G A366749-250MG A366749-5G
76648-574EA 36.08 USD
76648-574 76648-576 76648-578 76648-580
4-(3-Hydroxyazetidin-1-yl)benzoic acid 95%
4-(3-Hydroxyazetidin-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.2000 g/mol
MDL Number: MFCD26723054
CAS Number: 1416784-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR