Methyl 6-methoxy-1H-indazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A362902-1G A362902-250MG A362902-100MG
76648-572EA 636.54 USD
76648-572 77748-723 77748-722
Methyl 6-methoxy-1H-indazole-5-carboxylate 95%
Methyl 6-methoxy-1H-indazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.1980 g/mol
MDL Number: MFCD23705433
CAS Number: 1427405-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR