(2,2,2-Trifluoroethoxy)acetic acid 98%

Supplier: Ambeed
Danger

A360125-100MG A360125-250MG A360125-1G A360125-5G
76648-568EA 21 USD
76648-568 76648-570 77783-391 77783-392
(2,2,2-Trifluoroethoxy)acetic acid 98%
(2,2,2-Trifluoroethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₄H₅F₃O₃
MDL Number: MFCD02322998
CAS Number: 675-67-2
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR