2-Aminopyrimidine-4,6-diol 98%

Supplier: Ambeed
Warning

A359719-100G A359719-10G A359719-25G A359719-5G
76648-560EA 11.67 USD
76648-560 76648-562 76648-564 76648-566
2-Aminopyrimidine-4,6-diol 98%
2-Aminopyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₄H₅N₃O₂
MW: 127.1000 g/mol
MDL Number: MFCD00006094
CAS Number: 40497-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR