trans-3,5-dimethylmorpholine hydrochloride 97%

Supplier: Ambeed
Warning

A358192-100MG A358192-1G A358192-250MG
76648-546EA 40 USD
76648-546 76648-548 76648-550
trans-3,5-dimethylmorpholine hydrochloride 97%
trans-3,5-dimethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MDL Number: MFCD14525817
CAS Number: 2757083-05-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR