18-((2,5-Dioxopyrrolidin-1-yl)oxy)-18-oxooctadecanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
18-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-18-oxooctadecanoicacid
A357864-100MG
A357864-250MG
A357864-1G
A357864-5G
76648-542EA
128.37
USD
76648-542
76648-544
77748-711
77748-712
18-((2,5-Dioxopyrrolidin-1-yl)oxy)-18-oxooctadecanoic acid 95%
18-((2,5-Dioxopyrrolidin-1-yl)oxy)-18-oxooctadecanoic acid
Precaución: for Research Use Only
Formula:
C₂₂H₃₇NO₆ MW: 411.5323 g/mol |
MDL Number:
MFCD31698928 CAS Number: 146004-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...