3-Bromo-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A356723-100G A356723-1G A356723-25G A356723-5G A356723-10G
76648-534EA 156 USD
76648-534 76648-536 76648-538 76648-540 77572-048
3-Bromo-1H-1,2,4-triazole 98%
3-Bromo-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂H₂BrN₃
MW: 147.96 g/mol
MDL Number: MFCD01421960
CAS Number: 7343-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR