Benzenesulfinic chloride ≥95%

Supplier: Ambeed
Danger

A356294-1G A356294-5G
76648-522EA 141.1 USD
76648-522 76648-524
Benzenesulfinic chloride ≥95%
Benzenesulfinic chloride

Precaución: for Research Use Only

Formula: C₆H₅ClOS
MW: 160.6213 g/mol
MDL Number: MFCD01318288
CAS Number: 4972-29-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR