5,7-Dichloro-3-isopropyl-3H-imidazo[4,5-b]pyridine 95%

Supplier: Ambeed
Danger

A355773-100MG A355773-250MG
76648-518EA 180 USD
76648-518 76648-520
5,7-Dichloro-3-isopropyl-3H-imidazo[4,5-b]pyridine 95%
5,7-Dichloro-3-isopropyl-3H-imidazo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₉H₉Cl₂N₃
MW: 230.0939 g/mol
MDL Number: MFCD28402067
CAS Number: 1132747-05-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR