Potassium tri(1H-imidazol-1-yl)hydroborate 97%

Supplier: Ambeed
Warning

A355690-100MG A355690-1G A355690-5G A355690-250MG
76648-512EA 18 USD
76648-512 76648-514 76648-516 77622-292
Potassium tri(1H-imidazol-1-yl)hydroborate 97%
Potassium tri(1H-imidazol-1-yl)hydroborate

Precaución: for Research Use Only

Formula: C₉H₁₀BKN₆
CAS Number: 98047-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR