N1-Methyl-4-(trifluoromethoxy)benzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A352978-100MG A352978-250MG A352978-1G
76648-508EA 173.99 USD
76648-508 76648-510 77748-693
N1-Methyl-4-(trifluoromethoxy)benzene-1,2-diamine 95%
N1-Methyl-4-(trifluoromethoxy)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₉F₃N₂O
MW: 206.1700 g/mol
CAS Number: 1254166-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR